Assay Details
| Gene Symbol: Nrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497908.2 | XP_003497956.1 | 6 - 7 | 672 | 74 | |
| XM_007619380.1 | XP_007617570.1 | 7 - 8 | 778 | 74 | ||
| XM_007619388.1 | XP_007617578.1 | 7 - 8 | 772 | 74 | ||
| GenBank mRNA | JP039923.1 | - | 3 - 4 | 313 | 74 | |
More Information
Set Membership: |
|

Nrg4
Genomic Map