Assay Details
| Gene Symbol: Znf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498048.2 | XP_003498096.2 | 16 - 17 | 5187 | 61 | |
| XM_007621992.1 | XP_007620182.1 | 14 - 15 | 2829 | 61 | ||
| GenBank mRNA | JI889992.1 | - | 6 - 7 | 1412 | 61 | |
| JP056307.1 | - | 7 - 8 | 1510 | 61 | ||
More Information
Set Membership: |
|

Znf106
Genomic Map