Assay Details
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498217.2 | XP_003498265.1 | 7 - 8 | 726 | 61 | |
GenBank mRNA | JI882856.1 | - | 7 - 8 | 710 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
