Assay Details
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507874.2 | XP_003507922.2 | 11 - 12 | 1354 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507874.2 | XP_003507922.2 | 11 - 12 | 1354 | 56 |
Set Membership: |
![]() ![]() ![]() ![]() |