Assay Details
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511723.2 | XP_003511771.1 | 2 - 3 | 257 | 61 | |
| XM_007652257.1 | XP_007650447.1 | 2 - 3 | 257 | 61 | ||
More Information
Set Membership: |
|

Cacfd1
Genomic Map