Assay Details
| Gene Symbol: LOC100773268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512960.2 | XP_003513008.1 | 9 - 10 | 2071 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773268| Gene Symbol: LOC100773268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512960.2 | XP_003513008.1 | 9 - 10 | 2071 | 96 | |
Set Membership: |
|