Assay Details
| Gene Symbol: Pla2g4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513105.1 | XP_003513153.1 | 7 - 8 | 804 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g4c| Gene Symbol: Pla2g4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513105.1 | XP_003513153.1 | 7 - 8 | 804 | 64 | |
Set Membership: |
|