Assay Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514839.2 | XP_003514887.1 | 2 - 3 | 162 | 77 | |
| XM_007654883.1 | XP_007653073.1 | 2 - 3 | 378 | 77 | ||
| XM_007654884.1 | XP_007653074.1 | 3 - 4 | 517 | 77 | ||
| XM_007654885.1 | XP_007653075.1 | 2 - 3 | 163 | 77 | ||
More Information
Set Membership: |
|

Ccdc101
Genomic Map