Assay Details
| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650489.1 | XP_007648679.1 | 2 - 3 | 1500 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf10| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650489.1 | XP_007648679.1 | 2 - 3 | 1500 | 65 | |
Set Membership: |
|