Assay Details
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499609.1 | XP_003499657.1 | 3 - 4 | 256 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
