Assay Details
| Gene Symbol: LOC100771980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504129.2 | XP_003504177.1 | 6 - 7 | 620 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100771980| Gene Symbol: LOC100771980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504129.2 | XP_003504177.1 | 6 - 7 | 620 | 74 | |
Set Membership: |
|