Assay Details
| Gene Symbol: Glo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504453.2 | XP_003504501.1 | 3 - 4 | 416 | 66 | |
| GenBank mRNA | JI868341.1 | - | 3 - 4 | 395 | 66 | |
More Information
Set Membership: |
|

Glo1
Genomic Map