Assay Details
| Gene Symbol: Fam134c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504659.2 | XP_003504707.1 | 8 - 9 | 1036 | 92 | |
| GenBank mRNA | JI873681.1 | - | 1 - 2 | 113 | 92 | |
More Information
Set Membership: |
|

Fam134c
Genomic Map