Assay Details
| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504679.1 | XP_003504727.1 | 11 - 12 | 3936 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crb1| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504679.1 | XP_003504727.1 | 11 - 12 | 3936 | 58 | |
Set Membership: |
|