Assay Details
| Gene Symbol: Cttnbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515102.2 | XP_003515150.1 | 4 - 5 | 536 | 90 | |
| XM_007655105.1 | XP_007653295.1 | 5 - 6 | 742 | 90 | ||
More Information
Set Membership: |
|

Cttnbp2nl
Genomic Map