Assay Details
Gene Symbol: Ly6g5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649114.1 | XP_007647304.1 | 2 - 3 | 151 | 98 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ly6g5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649114.1 | XP_007647304.1 | 2 - 3 | 151 | 98 |