Assay Details
| Gene Symbol: Mrpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647363.1 | XP_007645553.1 | 6 - 7 | 838 | 60 | |
| GenBank mRNA | JP052566.1 | - | 6 - 7 | 838 | 60 | |
More Information
Set Membership: |
|

Mrpl38
Genomic Map