Assay Details
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650187.1 | XP_007648377.1 | 9 - 10 | 1506 | 93 | |
GenBank mRNA | JI882966.1 | - | 2 - 3 | 342 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
