Assay Details
| Gene Symbol: LOC100761290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647223.1 | XP_007645413.1 | 5 - 6 | 578 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100761290| Gene Symbol: LOC100761290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647223.1 | XP_007645413.1 | 5 - 6 | 578 | 66 | |
Set Membership: |
|