Assay Details
| Gene Symbol: Bhlhe40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498730.2 | XP_003498778.1 | 4 - 5 | 530 | 82 | |
| GenBank mRNA | JI864687.1 | - | 2 - 3 | 174 | 82 | |
| JP042839.1 | - | 2 - 3 | 174 | 82 | ||
More Information
Set Membership: |
|

Bhlhe40
Genomic Map