Assay Details
| Gene Symbol: LOC100751554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500837.2 | XP_003500885.1 | 4 - 5 | 471 | 109 | |
| GenBank mRNA | JP060618.1 | - | 4 - 5 | 474 | 109 | |
More Information
Set Membership: |
|

LOC100751554
Genomic Map