Assay Details
| Gene Symbol: Arpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502868.2 | XP_003502916.2 | 1 - 2 | 302 | 90 | |
| GenBank mRNA | JI879068.1 | - | 1 - 2 | 79 | 90 | |
More Information
Set Membership: |
|

Arpc5
Genomic Map