Assay Details
| Gene Symbol: Pir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503236.2 | XP_003503284.1 | 5 - 6 | 816 | 62 | |
| GenBank mRNA | JI866885.1 | - | 5 - 6 | 623 | 62 | |
More Information
Set Membership: |
|

Pir
Genomic Map