Assay Details
| Gene Symbol: Ppm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503263.2 | XP_003503311.1 | 3 - 4 | 886 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1l| Gene Symbol: Ppm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503263.2 | XP_003503311.1 | 3 - 4 | 886 | 67 | |
Set Membership: |
|