Assay Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514252.1 | XP_003514300.1 | 3 - 4 | 373 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1c2| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514252.1 | XP_003514300.1 | 3 - 4 | 373 | 61 | |
Set Membership: |
|