Assay Details
| Gene Symbol: Pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514978.2 | XP_003515026.2 | 10 - 11 | 1327 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnliprp2| Gene Symbol: Pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514978.2 | XP_003515026.2 | 10 - 11 | 1327 | 64 | |
Set Membership: |
|