Assay Details
| Gene Symbol: Calm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651153.1 | XP_007649343.1 | 4 - 5 | 462 | 76 | |
| GenBank mRNA | JI893250.1 | - | 4 - 5 | 462 | 76 | |
More Information
Set Membership: |
|

Calm3
Genomic Map