Assay Details
| Gene Symbol: LOC100758516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653915.1 | XP_007652105.1 | 11 - 12 | 1562 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758516| Gene Symbol: LOC100758516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653915.1 | XP_007652105.1 | 11 - 12 | 1562 | 102 | |
Set Membership: |
|