Assay Details
| Gene Symbol: LOC100753928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502764.2 | XP_003502812.1 | 2 - 3 | 167 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100753928| Gene Symbol: LOC100753928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502764.2 | XP_003502812.1 | 2 - 3 | 167 | 83 | |
Set Membership: |
|