Assay Details
| Gene Symbol: Ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506205.2 | XP_003506253.1 | 3 - 4 | 449 | 74 | |
| XM_007646720.1 | XP_007644910.1 | 3 - 4 | 485 | 74 | ||
More Information
Set Membership: |
|

Ormdl2
Genomic Map