Assay Details
| Gene Symbol: LOC100773442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508307.1 | XP_003508355.1 | 27 - 28 | 3850 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773442| Gene Symbol: LOC100773442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508307.1 | XP_003508355.1 | 27 - 28 | 3850 | 63 | |
Set Membership: |
|