Assay Details
| Gene Symbol: LOC100756666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508530.1 | XP_003508578.1 | 8 - 9 | 1142 | 78 | |
| XM_003508531.1 | XP_003508579.1 | 8 - 9 | 1142 | 78 | ||
| XM_007649150.1 | XP_007647340.1 | 8 - 9 | 1139 | 78 | ||
More Information
Set Membership: |
|

LOC100756666
Genomic Map