Assay Details
| Gene Symbol: Psd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648675.1 | XP_007646865.1 | 16 - 17 | 3035 | 64 | |
| XM_007648677.1 | XP_007646867.1 | 17 - 18 | 4221 | 64 | ||
| XM_007648678.1 | XP_007646868.1 | 16 - 17 | 3881 | 64 | ||
| XM_007648679.1 | XP_007646869.1 | 16 - 17 | 3032 | 64 | ||
More Information
Set Membership: |
|

Psd
Genomic Map