Assay Details
| Gene Symbol: Slamf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500272.1 | XP_003500320.1 | 1 - 2 | 78 | 76 | |
| XM_003500273.1 | XP_003500321.1 | 1 - 2 | 78 | 76 | ||
| XM_007640969.1 | XP_007639159.1 | 1 - 2 | 78 | 76 | ||
More Information
Set Membership: |
|

Slamf1
Genomic Map