Assay Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500359.2 | XP_003500407.1 | 4 - 5 | 2675 | 64 | |
| XM_007641142.1 | XP_007639332.1 | 4 - 5 | 1336 | 64 | ||
| XM_007641143.1 | XP_007639333.1 | 5 - 6 | 2458 | 64 | ||
More Information
Set Membership: |
|

Klhdc8b
Genomic Map