Assay Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504388.2 | XP_003504436.2 | 1 - 2 | 54 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp3| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504388.2 | XP_003504436.2 | 1 - 2 | 54 | 67 | |
Set Membership: |
|