Assay Details
| Gene Symbol: Bcl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515602.2 | XP_003515650.1 | 2 - 3 | 484 | 72 | |
| GenBank mRNA | JI890480.1 | - | 2 - 3 | 327 | 72 | |
| JP061652.1 | - | 2 - 3 | 327 | 72 | ||
More Information
Set Membership: |
|

Bcl10
Genomic Map