Assay Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646680.1 | XP_007644870.1 | 5 - 6 | 431 | 64 | |
| XM_007646681.1 | XP_007644871.1 | 5 - 6 | 392 | 64 | ||
| GenBank mRNA | JI884495.1 | - | 4 - 5 | 543 | 64 | |
More Information
Set Membership: |
|

Cs
Genomic Map