Assay Details
| Gene Symbol: Fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649191.1 | XP_007647381.1 | 1 - 2 | 344 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam155b| Gene Symbol: Fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649191.1 | XP_007647381.1 | 1 - 2 | 344 | 82 | |