Assay Details
| Gene Symbol: LOC100759746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650019.1 | XP_007648209.1 | 10 - 11 | 1391 | 100 | |
| XM_007650020.1 | XP_007648210.1 | 10 - 11 | 1391 | 100 | ||
More Information
Set Membership: |
|

LOC100759746
Genomic Map