Assay Details
| Gene Symbol: LOC100759746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650019.1 | XP_007648209.1 | 7 - 8 | 893 | 92 | |
| XM_007650020.1 | XP_007648210.1 | 7 - 8 | 893 | 92 | ||
| GenBank mRNA | JI887122.1 | - | 6 - 7 | 571 | 92 | |
More Information
Set Membership: |
|

LOC100759746
Genomic Map