Assay Details
| Gene Symbol: Znf184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652657.1 | XP_007650847.1 | 4 - 5 | 458 | 110 | |
| XM_007652658.1 | XP_007650848.1 | 5 - 6 | 625 | 110 | ||
| XM_007652659.1 | XP_007650849.1 | 5 - 6 | 686 | 110 | ||
| XM_007652660.1 | XP_007650850.1 | 2 - 3 | 168 | 110 | ||
| GenBank mRNA | JI876033.1 | - | 3 - 4 | 268 | 110 | |
| JP051394.1 | - | 3 - 4 | 268 | 110 | ||
More Information
Set Membership: |
|

Znf184
Genomic Map