Assay Details
| Gene Symbol: Actr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497612.2 | XP_003497660.1 | 10 - 11 | 1299 | 97 | |
| GenBank mRNA | JI872113.1 | - | 10 - 11 | 1254 | 97 | |
| JP037326.1 | - | 10 - 11 | 1254 | 97 | ||
More Information
Set Membership: |
|

Actr3
Genomic Map