Assay Details
| Gene Symbol: Ccdc122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500303.2 | XP_003500351.1 | 4 - 5 | 766 | 100 | |
| GenBank mRNA | JI862136.1 | - | 1 - 2 | 399 | 100 | |
| JP058607.1 | - | 4 - 5 | 749 | 100 | ||
More Information
Set Membership: |
|

Ccdc122
Genomic Map