Assay Details
| Gene Symbol: Ccdc74b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502842.2 | XP_003502890.1 | 1 - 2 | 129 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc74b| Gene Symbol: Ccdc74b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502842.2 | XP_003502890.1 | 1 - 2 | 129 | 98 | |