Assay Details
| Gene Symbol: Rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511319.2 | XP_003511367.1 | 3 - 4 | 414 | 94 | |
| XM_007651843.1 | XP_007650033.1 | 3 - 4 | 414 | 94 | ||
| XM_007651844.1 | XP_007650034.1 | 3 - 4 | 402 | 94 | ||
More Information
Set Membership: |
|

Rel
Genomic Map