Assay Details
| Gene Symbol: Taf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512482.2 | XP_003512530.1 | 2 - 3 | 135 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf13| Gene Symbol: Taf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512482.2 | XP_003512530.1 | 2 - 3 | 135 | 98 | |
Set Membership: |
|