Assay Details
| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515944.2 | XP_003515992.2 | 4 - 5 | 337 | 77 | |
| GenBank mRNA | JI876331.1 | - | 575 | 77 | ||
More Information
Set Membership: |
|

Lypla2
Genomic Map