Assay Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654689.1 | XP_007652879.1 | 15 - 16 | 1879 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Noc2l| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654689.1 | XP_007652879.1 | 15 - 16 | 1879 | 55 | |
Set Membership: |
|