Assay Details
| Gene Symbol: Rhou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653569.1 | XP_007651759.1 | 3 - 4 | 178 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhou| Gene Symbol: Rhou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653569.1 | XP_007651759.1 | 3 - 4 | 178 | 80 | |
Set Membership: |
|